Learn More
Waters Corp Waters Atlantis dC18 Column, 3 μm, 2.1 x 100 mm, 100 Å
Supplier:  Waters Corp 186001295
Encompass_Preferred
Atlantis dC18 columns are universal, silica-based, reversed-phase C18 columns designed for retaining polar compounds. These columns feature di-functionally bonded C18 ligands, optimized for use with highly aqueous mobile phases, including 100% water.
  • Universal, silica-based, reversed-phase C18 column
  • Designed for polar compound retention
  • Features di-functionally bonded C18 ligands
  • Optimized for use with highly aqueous mobile phases, including 100% water
  • Works well across the 3–7 pH range
Catalog No. 50-818-764