Learn More
Waters Corp Waters Column, 100Å, 3 μm, 2.1 mm × 150 mm
Catalog No. 50819057 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These universal, silica-based, reversed-phase C18 columns are designed for polar compound retention. They feature di-functionally bonded C18 ligands optimized for use with highly aqueous mobile phases, including 100% water. These columns offer an ideal balance of retention for both polar and non-polar compounds, are fully LC/MS-compatible, and provide superior retention of polar compounds without excessive retention of hydrophobic compounds compared to conventional reversed-phase HPLC columns.
  • Enhanced pH stability.
  • Compatible with aqueous mobile phases.
  • Superior peak shape.
  • Low pH stability.
  • Resists dewetting (hydrophobic collapse).
  • Enhanced polar compound retention.
  • Suitable for polar compound separation and standard reversed-phase applications.
  • Extended column lifetime and stability under acidic mobile conditions (due to difunctional silane bonding).
Catalog No. 50-819-057 Supplier Waters Corp Supplier No. 186001299
These universal, silica-based, reversed-phase C18 columns are designed for polar compound retention. They feature di-functionally bonded C18 ligands optimized for use with highly aqueous mobile phases, including 100% water. These columns offer an ideal balance of retention for both polar and non-polar compounds, are fully LC/MS-compatible, and provide superior retention of polar compounds without excessive retention of hydrophobic compounds compared to conventional reversed-phase HPLC columns.
  • Enhanced pH stability.
  • Compatible with aqueous mobile phases.
  • Superior peak shape.
  • Low pH stability.
  • Resists dewetting (hydrophobic collapse).
  • Enhanced polar compound retention.
  • Suitable for polar compound separation and standard reversed-phase applications.
  • Extended column lifetime and stability under acidic mobile conditions (due to difunctional silane bonding).