Learn More
Waters Corp Waters Corporation Atlantis dC18 Column, 5 μm, 2.1 mm × 150 mm
Supplier:  Waters Corp 186001301
Encompass_Preferred
Atlantis dC18 columns are universal, silica-based, reversed-phase C18 columns used for polar compound retention. They feature di-functionally bonded C18 ligands optimized for use with highly aqueous mobile phases, including 100% water.
  • Designed for polar compound retention and enhanced retention of polar analytes.
  • Features di-functionally bonded C18 ligands for highly aqueous mobile phases, including 100% water.
  • Counteracts “dewetting” for effective operation in aqueous mobile phases.
  • 100Å pore size suitable for small to medium-sized polar compounds.
  • Suitable for reversed-phase chromatography of polar and non-polar compounds across various applications.
  • Offers high pH stability for improved separation of certain analytes.
  • Requires flushing with water and organic solvents post-analysis and proper storage.
Catalog No. 50-818-610