Learn More
Waters Corp Atlantis dC18 Column, 5 μm, 100 Å, 4.6 mm × 150 mm
Catalog No. 50818674 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Atlantis dC18 columns are universal, silica-based, reversed-phase C18 columns for polar compound retention. They feature di-functionally bonded C18 ligands, optimized for highly aqueous mobile phases, including 100% water. These columns ensure superior peak shape, low pH stability, and resistance to dewetting. They are fully end-capped for compatibility with aqueous mobile phases and full LC/MS compatibility.
  • Universal, silica-based, reversed-phase C18 chemistry.
  • Optimized for polar compound retention and highly aqueous mobile phases.
  • Superior peak shape, low pH stability, and dewetting resistance.
  • Fully end-capped for desirable peak shape with bases.
  • Full LC/MS compatibility.
  • Manufactured in a cGMP, ISO 9001:200 certified plant.
Catalog No. 50-818-674 Supplier Waters Corp Supplier No. 186001344
Atlantis dC18 columns are universal, silica-based, reversed-phase C18 columns for polar compound retention. They feature di-functionally bonded C18 ligands, optimized for highly aqueous mobile phases, including 100% water. These columns ensure superior peak shape, low pH stability, and resistance to dewetting. They are fully end-capped for compatibility with aqueous mobile phases and full LC/MS compatibility.
  • Universal, silica-based, reversed-phase C18 chemistry.
  • Optimized for polar compound retention and highly aqueous mobile phases.
  • Superior peak shape, low pH stability, and dewetting resistance.
  • Fully end-capped for desirable peak shape with bases.
  • Full LC/MS compatibility.
  • Manufactured in a cGMP, ISO 9001:200 certified plant.