Learn More
Waters Corp Atlantis Sentry Guard Cartridge, dC18, 100Å, 5 μm, 4.6 mm x 20 mm
Catalog No. 50819125 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Atlantis dC18 Sentry Guard Cartridge extends analytical column life by eliminating chemical and particle pollution from the mobile phase stream. It protects 4.6 mm I.D. cartridges and works with Atlantis dC18 Sentry Columns for reversed-phase Liquid Chromatography.
  • Designed to extend analytical column life
  • Eliminates chemical and particle pollution
  • Protects 4.6 mm I.D. cartridges
  • Made with same packing materials as analytical column for consistent performance
  • Helps UPLC analytical columns last longer
  • Removes contaminants while maintaining separation efficiency in UPLC, UHPLC, and HPLC
  • Manufactured under strict cGMP, ISO 9001 conditions
  • Tested and certified for optimal performance
  • Improves selectivity, sensitivity, and speed of analysis
Catalog No. 50-819-125 Supplier Waters Corp Supplier No. 186001323
The Atlantis dC18 Sentry Guard Cartridge extends analytical column life by eliminating chemical and particle pollution from the mobile phase stream. It protects 4.6 mm I.D. cartridges and works with Atlantis dC18 Sentry Columns for reversed-phase Liquid Chromatography.
  • Designed to extend analytical column life
  • Eliminates chemical and particle pollution
  • Protects 4.6 mm I.D. cartridges
  • Made with same packing materials as analytical column for consistent performance
  • Helps UPLC analytical columns last longer
  • Removes contaminants while maintaining separation efficiency in UPLC, UHPLC, and HPLC
  • Manufactured under strict cGMP, ISO 9001 conditions
  • Tested and certified for optimal performance
  • Improves selectivity, sensitivity, and speed of analysis