Learn More
Waters Corp Prep Guard Cartridge, dC18, 10 μm, 19 mm x 10 mm
Catalog No. 50780673 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This guard cartridge is designed for the retention and separation of polar analytes in chromatography. Featuring dC18 chemistry, it offers superior retention of polar compounds and is compatible with 100% aqueous mobile phases. It delivers exceptional reproducible performance for both polar and non-polar molecules.
  • Designed for reproducible retention and separation of polar and non-polar compounds
  • Compatible with 100% aqueous mobile phases
  • Features tri-functional C18 bonding chemistry
  • Achieves long column lifetimes even with low-pH mobile phases
  • Offers analytical column performance in preparative chromatography formats
  • Provides improved bed stability, higher efficiency, and increased loadability
Catalog No. 50-780-673 Supplier Waters Corp Supplier No. 186001363
This guard cartridge is designed for the retention and separation of polar analytes in chromatography. Featuring dC18 chemistry, it offers superior retention of polar compounds and is compatible with 100% aqueous mobile phases. It delivers exceptional reproducible performance for both polar and non-polar molecules.
  • Designed for reproducible retention and separation of polar and non-polar compounds
  • Compatible with 100% aqueous mobile phases
  • Features tri-functional C18 bonding chemistry
  • Achieves long column lifetimes even with low-pH mobile phases
  • Offers analytical column performance in preparative chromatography formats
  • Provides improved bed stability, higher efficiency, and increased loadability