Learn More
Waters Corp Atlantis Column, C18, 3 μm, 100 Å, 4.6 mm × 150 mm
Catalog No. 502391075 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This column features an electronic eConnect column tag that contains unique column-specific information. These NFC-enabled, device-tagged columns were designed to automatically identify, track, and digitally trace the column's use history. While eConnect columns can be utilized with any HPLC system, their full advantages are realized when paired with the Waters Alliance iS System running Empower software.
  • Electronic eConnect column tag for automatic tracking and digital tracing.
  • Compatible with any HPLC system.
  • C18 chemistry for reversed phase separation.
  • Silica particle substrate, spherical shape, 3 μm particle size.
  • 100 Å pore size.
  • pH range of 3 to 7.
  • Maximum pressure of 6000 psi (415 Bar).
  • Endcapped with dC18 bonding technology.
  • 4.6 mm inner diameter and 150 mm length.
  • Suitable for small molecule applications.
Catalog No. 50-239-1075 Supplier Waters Corp Supplier No. 186001342RF
This column features an electronic eConnect column tag that contains unique column-specific information. These NFC-enabled, device-tagged columns were designed to automatically identify, track, and digitally trace the column's use history. While eConnect columns can be utilized with any HPLC system, their full advantages are realized when paired with the Waters Alliance iS System running Empower software.
  • Electronic eConnect column tag for automatic tracking and digital tracing.
  • Compatible with any HPLC system.
  • C18 chemistry for reversed phase separation.
  • Silica particle substrate, spherical shape, 3 μm particle size.
  • 100 Å pore size.
  • pH range of 3 to 7.
  • Maximum pressure of 6000 psi (415 Bar).
  • Endcapped with dC18 bonding technology.
  • 4.6 mm inner diameter and 150 mm length.
  • Suitable for small molecule applications.