Learn More
Waters Corp Column, C18, 3 μm, 100 Å, 4.6 mm x 100 mm
Supplier:  Waters Corp 186001337RF
Encompass_Preferred
The Atlantis dC18 Column is a universal, silica-based, reversed-phase C18 column, optimized for retaining polar compounds even with 100% aqueous mobile phases. It balances retention for polar and nonpolar compounds, is LC/MS compatible, and offers enhanced pH stability.
  • Ideal balance of retention for polar and nonpolar compounds.
  • Fully LC/MS compatible.
  • Enhanced pH stability from difunctional silane bonding chemistry.
  • Exceptional peak shapes for bases with full end-capping.
  • Suitable for polar compound separation and standard reversed-phase applications.
  • Extended column lifetime guaranteed.
Catalog No. 50-239-1881