Learn More
Waters Corp Atlantis dC18 Column, 100 Å, 5 μm, 4.6 mm x 150 mm
Catalog No. 502390523 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This column is equipped with an electronic eConnect column tag that contains unique, column-specific information. These NFC-enabled device-tagged columns are designed for automatic identification, tracking, and digital tracing of column use history. While eConnect columns can be used on any HPLC system, their advantages are fully realized when used with the Waters Alliance iS System running Empower software.
  • Equipped with an electronic eConnect column tag for unique column-specific information.
  • NFC-enabled device-tagged for automatic identification, tracking, and digital tracing of column use history.
  • Can be used on any HPLC system.
  • Advantages are unlocked when used with the Alliance iS System running Empower software.
Catalog No. 50-239-0523 Supplier Waters Corp Supplier No. 186001344RF
Add to Cart
Add to Cart
This column is equipped with an electronic eConnect column tag that contains unique, column-specific information. These NFC-enabled device-tagged columns are designed for automatic identification, tracking, and digital tracing of column use history. While eConnect columns can be used on any HPLC system, their advantages are fully realized when used with the Waters Alliance iS System running Empower software.
  • Equipped with an electronic eConnect column tag for unique column-specific information.
  • NFC-enabled device-tagged for automatic identification, tracking, and digital tracing of column use history.
  • Can be used on any HPLC system.
  • Advantages are unlocked when used with the Alliance iS System running Empower software.