Learn More
Waters Corp Waters Corporation Column, C18, 3 μm, 3.9 mm × 150 mm
Catalog No. 50780657 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This universal, silica-based, reversed-phase C18 column is designed for the retention of polar compounds. It features di-functionally bonded C18 ligands optimized for highly aqueous mobile phases, including 100% water. The column offers superior peak shape, low pH stability, resistance to dewetting, and enhanced polar compound retention.
  • Superior peak shape
  • Low pH stability
  • Resistance to dewetting
  • Enhanced polar compound retention
  • Fully LC/MS-compatible
  • Suitable for separating both polar compounds and for standard reversed-phase applications
  • End-capped to avoid peak tailing and extreme retention for amine-containing bases
  • Optimal ligand density allows strong retention of polar compounds without excessive retention of non-polar compounds
Catalog No. 50-780-657 Supplier Waters Corp Supplier No. 186001317
This universal, silica-based, reversed-phase C18 column is designed for the retention of polar compounds. It features di-functionally bonded C18 ligands optimized for highly aqueous mobile phases, including 100% water. The column offers superior peak shape, low pH stability, resistance to dewetting, and enhanced polar compound retention.
  • Superior peak shape
  • Low pH stability
  • Resistance to dewetting
  • Enhanced polar compound retention
  • Fully LC/MS-compatible
  • Suitable for separating both polar compounds and for standard reversed-phase applications
  • End-capped to avoid peak tailing and extreme retention for amine-containing bases
  • Optimal ligand density allows strong retention of polar compounds without excessive retention of non-polar compounds