Learn More
Waters Corp Waters Atlantis dC18 Column, 100Å, 3 μm, 4.6 mm x 100 mm
Catalog No. 50780667 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Atlantis dC18 columns are universal, silica-based, reversed-phase C18 columns designed for polar compound retention. They are optimized for use with highly aqueous mobile phases, including 100% water, and offer an ideal balance of retention for both polar and nonpolar compounds.
  • Features di-functionally bonded C18 ligands.
  • Fully LC/MS compatible.
  • Delivers superior retention of polar compounds.
  • Offers enhanced pH stability with aqueous mobile phases.
  • Packing material created for separation of polar, water-soluble compounds.
  • Combines optimized stationary phase attributes for superior peak shape and low pH stability.
  • Guarantees extended column lifetime.
  • Comes packed and shipped in 100% acetonitrile.
  • Can be equilibrated using 10 column volumes of the mobile phase.
Catalog No. 50-780-667 Supplier Waters Corp Supplier No. 186001337
Atlantis dC18 columns are universal, silica-based, reversed-phase C18 columns designed for polar compound retention. They are optimized for use with highly aqueous mobile phases, including 100% water, and offer an ideal balance of retention for both polar and nonpolar compounds.
  • Features di-functionally bonded C18 ligands.
  • Fully LC/MS compatible.
  • Delivers superior retention of polar compounds.
  • Offers enhanced pH stability with aqueous mobile phases.
  • Packing material created for separation of polar, water-soluble compounds.
  • Combines optimized stationary phase attributes for superior peak shape and low pH stability.
  • Guarantees extended column lifetime.
  • Comes packed and shipped in 100% acetonitrile.
  • Can be equilibrated using 10 column volumes of the mobile phase.