Learn More
Waters Corp Waters Atlantis dC18 Column, 5 μm, 100 Å, 2.1 mm, 100 mm
Catalog No. 50780647 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Atlantis dC18 columns are universal, silica-based, reversed-phase C18 columns designed for polar compound retention. They utilize di-functionally bonded C18 ligands optimized for highly aqueous mobile phases, including 100% water. Suitable for both LC and MS, these columns balance retention for polar and nonpolar molecules while limiting hydrophobic substance retention. They offer improved pH stability and compatibility with water mobile phases, enabling effective separation of polar, water-soluble compounds in reversed-phase HPLC.
  • Universal C18 analytical column for LC and MS applications
  • Optimized for highly aqueous mobile phases, including 100% water
  • Provides enhanced peak shape, low pH stability, improved polar compound retention, and resistance to hydrophobic collapse
  • Di-functional silane bonding chemistry ensures good stability under acidic mobile conditions and extended column lifespan
Catalog No. 50-780-647 Supplier Waters Corp Supplier No. 186001297
Add to Cart
Add to Cart
Atlantis dC18 columns are universal, silica-based, reversed-phase C18 columns designed for polar compound retention. They utilize di-functionally bonded C18 ligands optimized for highly aqueous mobile phases, including 100% water. Suitable for both LC and MS, these columns balance retention for polar and nonpolar molecules while limiting hydrophobic substance retention. They offer improved pH stability and compatibility with water mobile phases, enabling effective separation of polar, water-soluble compounds in reversed-phase HPLC.
  • Universal C18 analytical column for LC and MS applications
  • Optimized for highly aqueous mobile phases, including 100% water
  • Provides enhanced peak shape, low pH stability, improved polar compound retention, and resistance to hydrophobic collapse
  • Di-functional silane bonding chemistry ensures good stability under acidic mobile conditions and extended column lifespan