Learn More
Waters Corp Waters Atlantis dC18 Column, 100Å, 5 μm, 3.9 mm × 150 mm
Supplier:  Waters Corp 186001319
Encompass_Preferred
Atlantis dC18 columns are universal, silica-based, reversed-phase C18 columns. They are designed for polar compound retention, compatible with both LC and MS systems, and optimized for highly aqueous mobile phases including 100% water. They offer improved pH stability.
  • Balances retention for polar and nonpolar chemicals.
  • Minimizes hydrophobic compound retention.
  • Features silica-based, di-functionally bonded C18 material.
  • Optimized stationary phase delivers superior peak form.
  • Resists hydrophobic collapse.
  • Di-functional silane bonding chemistry extends column lifetime.
Catalog No. 50-780-658