Learn More
Waters Corp Waters Atlantis dC18 Column, 5 μm, 3 mm, 150 mm, 100 Å
Supplier:  Waters Corp 186001309
Encompass_Preferred
Atlantis dC18 columns are universal, silica-based, reversed-phase C18 columns for polar compound retention, featuring di-functionally bonded C18 ligands optimized for highly aqueous mobile phases, including 100% water. They are fully compatible with LC/MS and engineered for balanced retention of polar and nonpolar chemicals.
  • Improved pH stability.
  • Provides superior peak form.
  • Resistance to hydrophobic collapse (dewetting).
  • Good stability under acidic mobile conditions.
  • Longer column lifetime.
  • Can tolerate operating pressures up to 6000 psi (415 Bar) and a pH range of 3 to 7.
Catalog No. 50-780-653