Learn More
Waters Corp Waters Corporation Atlantis dC18 OBD Column, 5 μm, 30 x 50 mm
Supplier:  Waters Corp 186001373
Encompass_Preferred
Atlantis dC18 OBD Prep Columns feature dC18, 100 Å pore size technology and are designed for fast, efficient, lab-scale separations for greater throughput.
  • Achieve fast, efficient, lab-scale separations for greater throughput.
  • Directly scale from UPLC, UHPLC, or HPLC screening to lab-scale purification.
  • Select robust chromatographic particles designed for purification.
  • Provide column stability and reliability with long, predictable lifetimes, showing exceptional resistance to mechanical chromatographic bed failure.
  • Ensure scalability from analytical to preparative dimensions by matching column bed densities.
Catalog No. 50-780-678