Learn More
Waters Corp Waters Atlantis dC18 Column, 100Å, 5 μm, 4.6 mm × 100 mm
Catalog No. 50780668 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Atlantis dC18 columns are universal, silica-based, reversed-phase C18 columns used for polar compound retention. They feature di-functionally bonded C18 ligands optimized for use with highly aqueous mobile phases, including 100% water. This analytical column is compatible with both LC and MS, designed to provide an excellent balance of retention for polar and nonpolar molecules while limiting retention for hydrophobic substances. The column also offers improved pH stability, making it suitable for water mobile phases.
  • Enhanced peak shape.
  • Low pH stability.
  • Improved polar compound retention.
  • Resistance to hydrophobic collapse.
  • Good stability under acidic mobile conditions.
  • Longer column lifespan.
  • Suitable for both polar compound separation and common reversed-phase applications.
Catalog No. 50-780-668 Supplier Waters Corp Supplier No. 186001340
Add to Cart
Add to Cart
Atlantis dC18 columns are universal, silica-based, reversed-phase C18 columns used for polar compound retention. They feature di-functionally bonded C18 ligands optimized for use with highly aqueous mobile phases, including 100% water. This analytical column is compatible with both LC and MS, designed to provide an excellent balance of retention for polar and nonpolar molecules while limiting retention for hydrophobic substances. The column also offers improved pH stability, making it suitable for water mobile phases.
  • Enhanced peak shape.
  • Low pH stability.
  • Improved polar compound retention.
  • Resistance to hydrophobic collapse.
  • Good stability under acidic mobile conditions.
  • Longer column lifespan.
  • Suitable for both polar compound separation and common reversed-phase applications.