Learn More
Waters Corp Waters Column, dC18, 5 μm, 4.6 mm × 75 mm, 100 Å
Supplier:  Waters Corp 186001335
Encompass_Preferred
Atlantis dC18 columns are universal, silica-based, reversed-phase C18 columns for polar compound retention. They use di-functionally bonded C18 ligands optimized for highly aqueous mobile phases, including 100% water, delivering superb peak shapes for bases.
  • Universal, silica-based, reversed-phase C18 column designed for polar compound retention.
  • Di-functionally bonded C18 ligands optimized for highly aqueous mobile phases, including 100% water.
  • Delivers superb peak shapes for bases.
  • Fully end-capped.
  • Advanced stationary phase compatible with aqueous mobile phases for retaining hydrophilic compounds.
  • LC/MS compatible.
  • Offers greater retention of polar molecules compared to traditional reversed-phase HPLC columns without excessive retention of hydrophobic chemicals.
  • Improved pH stability.
Catalog No. 50-780-666