Learn More
Waters Corp Atlantis dC18 IS Column, 3 μm, 4.6 mm x 20 mm
Supplier: Waters Corp 186002062
The Atlantis dC18 Intelligent Speed (IS) Column is engineered for high-performance liquid chromatography (HPLC) applications, utilizing C18 chemistry on a spherical silica particle substrate. This column enables efficient reversed-phase separations, providing excellent efficiency and fast analysis.
- C18 chemistry, reversed phase separation.
- Spherical silica particle substrate.
- pH range: 3 to 7.
- Maximum pressure: 6000 psi (415 Bar).
- Endcapped dC18 bonding technology.
- Medium silanol activity.
- Particle size: 3 μm; pore size: 100 Å.
- Dimensions: 4.6 mm ID x 20 mm L.
- 12% carbon load.
- Intelligent Speed (IS) technology for faster, efficient analysis.
- Applications: peptide, protein, small molecule analysis, drug discovery.
- Benefits: excellent separation, fast analysis, versatility, durability.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.