Learn More
Zymo Research Corporation Atlantis dsDNase 12.5 U (0.1 U/ul)
SDP
Catalog No. 50444531
Encompass_Preferred
Atlantis dsDNase is a double-strand DNA specific endonuclease that cleaves phosphodiester bonds in DNA to yield homogeneous populations of core nucleosomes.
Catalog No. 50-444-531 Supplier Zymo Research Corporation Supplier No. E2030
Atlantis dsDNase is a double-strand DNA specific endonuclease that cleaves phosphodiester bonds in DNA to yield homogeneous populations of core nucleosomes.