Learn More
Waters Corp Waters HILIC Column, Silica, 3 μm, 2.1 mm, 50 mm
Supplier: Waters Corp 186002011
Atlantis HILIC Silica columns are designed for use with high organic mobile phases (>80% acetonitrile) to retain analytes too polar for reverse-phase chromatography. They effectively separate very polar, water-soluble basic organic compounds, including peptides, metabolites, and actives, leading to higher sensitivity and lower limits of detection when combined with ESI-MS.
- HILIC chemistry
- Hydrophilic Interaction (HILIC) separation mode
- Silica particle substrate
- pH range: 1 to 5 pH
- Maximum pressure: 6000 psi (415 Bar)
- Not endcapped
- High silanol activity
- Spherical particle shape
- 3 μm particle size
- 100 Å pore size
- Column format
- 330 surface area
- HPLC system compatibility
- Silica particle technology
- L3 USP classification
- 2.1 mm inner diameter
- 50 mm length
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.