Learn More
Waters Corp Waters Atlantis HILIC Silica Column, 3 μm, 100 Å, 3 mm x 50 mm
Catalog No. 50819130 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Atlantis HILIC Silica columns are designed for use with high organic mobile phases (>80% acetonitrile) to retain analytes that are too polar for reverse-phase chromatography.
  • Retains highly polar molecules.
  • Improves LC/ESI-MS response.
  • Direct compatibility with SPE solvents.
  • Offers complementary selectivity to reversed-phase HPLC.
  • Suitable for metabolism, drug discovery, and combinatorial chemistry.
  • Operates within 1 to 5 pH range.
  • Withstands 6,000 psi (415 Bar) maximum pressure.
Catalog No. 50-819-130 Supplier Waters Corp Supplier No. 186002017
Atlantis HILIC Silica columns are designed for use with high organic mobile phases (>80% acetonitrile) to retain analytes that are too polar for reverse-phase chromatography.
  • Retains highly polar molecules.
  • Improves LC/ESI-MS response.
  • Direct compatibility with SPE solvents.
  • Offers complementary selectivity to reversed-phase HPLC.
  • Suitable for metabolism, drug discovery, and combinatorial chemistry.
  • Operates within 1 to 5 pH range.
  • Withstands 6,000 psi (415 Bar) maximum pressure.