Learn More
Waters Corp Atlantis Column, HILIC, Silica, 100 Å Pore, 3 μm Particle
Catalog No. 50780885 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Atlantis HILIC Silica columns retain polar analytes when used with high organic mobile phases, offering orthogonal selectivity to reverse-phase chromatography. Ideal for metabolism, drug discovery, and combinatorial chemistries, especially for basic polar analytes.
  • Retain peptides, metabolites, and actives; improved LC/ESI-MS response; direct compatibility with SPE solvents.
  • Complementary selectivity to reversed-phase HPLC; retains highly polar basic analytes with volatile mobile phases for higher sensitivity in ESI-MS.
  • Useful in drug metabolism studies for polar, low-concentration metabolites.
  • Long lifetime, universal compatibility with all LC detectors, and outstanding column-to-column reproducibility.
  • Superior peak shapes for polar bases, facilitating faster and easier method development.
Catalog No. 50-780-885 Supplier Waters Corp Supplier No. 186002029
Add to Cart
Add to Cart
Atlantis HILIC Silica columns retain polar analytes when used with high organic mobile phases, offering orthogonal selectivity to reverse-phase chromatography. Ideal for metabolism, drug discovery, and combinatorial chemistries, especially for basic polar analytes.
  • Retain peptides, metabolites, and actives; improved LC/ESI-MS response; direct compatibility with SPE solvents.
  • Complementary selectivity to reversed-phase HPLC; retains highly polar basic analytes with volatile mobile phases for higher sensitivity in ESI-MS.
  • Useful in drug metabolism studies for polar, low-concentration metabolites.
  • Long lifetime, universal compatibility with all LC detectors, and outstanding column-to-column reproducibility.
  • Superior peak shapes for polar bases, facilitating faster and easier method development.