Learn More
Waters Corp Waters Atlantis HILIC Column, 100Å, 5 μm, 2.1 mm × 150 mm
Catalog No. 50780872 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These HILIC Silica columns are designed for use with high organic mobile phases (>80% acetonitrile) to retain analytes that are too polar for reverse-phase chromatography. This separation mechanism provides orthogonal analyte selectivity, enabling retention of very polar, water-soluble basic organic compounds. They are useful for processing peptides, metabolites, and actives, offering improved LC/ESI-MS response and direct compatibility with SPE solvents.
  • Retains analytes too polar for reverse-phase chromatography.
  • Provides orthogonal analyte selectivity.
  • Enables separation of very polar, water-soluble basic organic compounds.
  • Offers improved LC/ESI-MS response.
  • Delivers higher sensitivity and lower limits of detection.
  • Provides long column lifetime and universal compatibility.
  • Achieves superior peak shapes for polar bases.
Catalog No. 50-780-872 Supplier Waters Corp Supplier No. 186002016
These HILIC Silica columns are designed for use with high organic mobile phases (>80% acetonitrile) to retain analytes that are too polar for reverse-phase chromatography. This separation mechanism provides orthogonal analyte selectivity, enabling retention of very polar, water-soluble basic organic compounds. They are useful for processing peptides, metabolites, and actives, offering improved LC/ESI-MS response and direct compatibility with SPE solvents.
  • Retains analytes too polar for reverse-phase chromatography.
  • Provides orthogonal analyte selectivity.
  • Enables separation of very polar, water-soluble basic organic compounds.
  • Offers improved LC/ESI-MS response.
  • Delivers higher sensitivity and lower limits of detection.
  • Provides long column lifetime and universal compatibility.
  • Achieves superior peak shapes for polar bases.