Learn More
Waters Corp Atlantis HILIC Column, Silica, 3 μm, 100 Å, 4.6 × 50 mm
Catalog No. 50780883 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Atlantis HILIC Silica columns efficiently separate very polar compounds not retained by reverse-phase chromatography. They utilize high organic mobile phases and Hydrophilic Interaction Chromatography (HILIC) for orthogonal selectivity. These columns enhance LC/ESI-MS response and are compatible with SPE solvents, leading to higher sensitivity.
  • Exceptional retention of very polar compounds.
  • Improved LC/ESI-MS response.
  • Direct compatibility with SPE solvents, enhancing sensitivity and lowering detection limits.
  • Complementary selectivity to reversed-phase HPLC columns for challenging basic polar analytes.
  • Long column lifetime.
  • Outstanding column-to-column reproducibility.
Catalog No. 50-780-883 Supplier Waters Corp Supplier No. 186002027
Atlantis HILIC Silica columns efficiently separate very polar compounds not retained by reverse-phase chromatography. They utilize high organic mobile phases and Hydrophilic Interaction Chromatography (HILIC) for orthogonal selectivity. These columns enhance LC/ESI-MS response and are compatible with SPE solvents, leading to higher sensitivity.
  • Exceptional retention of very polar compounds.
  • Improved LC/ESI-MS response.
  • Direct compatibility with SPE solvents, enhancing sensitivity and lowering detection limits.
  • Complementary selectivity to reversed-phase HPLC columns for challenging basic polar analytes.
  • Long column lifetime.
  • Outstanding column-to-column reproducibility.