Learn More
Waters Corp HILIC Column, 5 μm, 100 Å, 4.6 mm x 50 mm
Supplier: Waters Corp 186002028
Atlantis HILIC Silica Columns are specifically designed and tested for Hydrophilic Interaction Chromatography (HILIC) separations. They are excellent for the retention of very polar, basic, water-soluble analytes and employ highly volatile (>80% organic) mobile phases, making them ideal for mass spectrometry (MS) response and sensitivity. These columns retain very polar, organic molecules that cannot be retained using reversed-phase (RP) methods.
- Excellent for retention of very polar, basic, water-soluble analytes.
- Designed for HILIC separations.
- Employs highly volatile (>80% organic) mobile phases.
- Ideal for mass spectrometry (MS) response and sensitivity.
- Ideal for diverse analyte mixtures.
- Manufactured using ultrapure reagents in a cGMP, ISO 9001:2000 certified facility.
- Individually tested with Certificate of Batch Analysis and Performance Chromatogram.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.