Learn More
Waters Corp Waters Atlantis HILIC Column, Silica, 100Å, 3 μm, 2.1 mm × 30 mm
Supplier:  Waters Corp 186002009
Encompass_Preferred
These HILIC Silica columns utilize high organic mobile phases (>80% acetonitrile) to retain analytes too polar for reverse-phase chromatography, offering orthogonal selectivity. They effectively separate and retain highly polar, water-soluble basic chemical molecules, enhancing LC/ESI-MS performance and providing compatibility with SPE solvents.
  • Designed for high organic mobile phases (>80% acetonitrile).
  • Retains analytes too polar for reversed-phase chromatography.
  • Provides orthogonal analyte selectivity.
  • Separates highly polar, water-soluble basic chemical molecules.
  • Enhances LC/ESI-MS performance.
  • Offers direct compatibility with SPE solvents.
  • Suitable for metabolism, drug discovery, and combinatorial chemistry applications.
  • Enables higher sensitivity and lower detection limits.
Catalog No. 50-780-865