Learn More
Waters Corp Atlantis HILIC Column, 3 μm, 100Å, 3 mm × 100 mm
Supplier:  Waters Corp 186002019
Encompass_Preferred
Atlantis HILIC Silica columns are designed for high organic mobile phases (>80% acetonitrile) to retain analytes too polar for reverse-phase chromatography, offering orthogonal analyte selectivity compared to traditional reversed-phase methods.
  • Provides orthogonal analyte selectivity to traditional reversed-phase approaches.
  • Utilizes HILIC technology for compound elution ordered by increasing hydrophilicity.
  • Partitions polar analytes between water-rich and water-poor mobile phases.
  • High-quality silica stationary phases create weak cation exchanges for polar bases.
  • Enables retention for highly polar basic analytes with a net positive charge at acidic pHs.
  • Offers extremely high ESI-MS sensitivity and lower limits of detection.
  • Increases throughput and reduces time for sample preparation techniques.
Catalog No. 50-780-875