Learn More
Waters Corp Waters HILIC Column, 100Å, 3 μm, 2.1 mm × 150 mm
Catalog No. 50780871 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Atlantis HILIC Silica columns are designed for use with high organic mobile phases (>80% acetonitrile) to retain analytes that are too polar for reverse-phase chromatography. The HILIC separation mechanisms offer orthogonal analyte selectivity compared to traditional reversed-phase chromatographic approaches.
  • HILIC chemistry
  • Hydrophilic interaction (HILIC) separation mode
  • Silica particle substrate
  • pH range of 1 to 5
  • Maximum pressure of 6000 psi (415 bar)
  • Not endcapped
  • High silanol activity
  • Spherical particle shape
  • 3 μm particle size
  • 100 Å pore size
  • Column format
  • Surface area of 330
  • HPLC system compatible
  • Silica particle technology
  • L3 USP classification
  • 2.1 mm inner diameter
  • 150 mm length
  • 1 unit per package
Catalog No. 50-780-871 Supplier Waters Corp Supplier No. 186002015
Atlantis HILIC Silica columns are designed for use with high organic mobile phases (>80% acetonitrile) to retain analytes that are too polar for reverse-phase chromatography. The HILIC separation mechanisms offer orthogonal analyte selectivity compared to traditional reversed-phase chromatographic approaches.
  • HILIC chemistry
  • Hydrophilic interaction (HILIC) separation mode
  • Silica particle substrate
  • pH range of 1 to 5
  • Maximum pressure of 6000 psi (415 bar)
  • Not endcapped
  • High silanol activity
  • Spherical particle shape
  • 3 μm particle size
  • 100 Å pore size
  • Column format
  • Surface area of 330
  • HPLC system compatible
  • Silica particle technology
  • L3 USP classification
  • 2.1 mm inner diameter
  • 150 mm length
  • 1 unit per package