Learn More
Waters Corp Waters HILIC Column, 100Å, 5 μm, 4.6 mm X 150 mm
Catalog No. 50780888 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Atlantis HILIC Silica columns retain analytes too polar for reverse-phase chromatography, especially with high organic mobile phases (>80% acetonitrile). This HILIC separation offers orthogonal selectivity for separating water-soluble, basic, polar organic molecules such as peptides, metabolites, and actives.
  • Provides better LC/ESI-MS response than reversed-phase HPLC columns.
  • Offers direct compatibility with SPE solvents.
  • Delivers complementary selectivity.
  • Achieves higher sensitivity and lower detection limits in drug metabolism research.
  • Ensures universal compatibility with all LC detectors.
  • Guarantees excellent column-to-column repeatability.
  • Improves peak morphologies for polar bases.
  • Facilitates faster and easier method development.
Catalog No. 50-780-888 Supplier Waters Corp Supplier No. 186002032
Add to Cart
Add to Cart
Atlantis HILIC Silica columns retain analytes too polar for reverse-phase chromatography, especially with high organic mobile phases (>80% acetonitrile). This HILIC separation offers orthogonal selectivity for separating water-soluble, basic, polar organic molecules such as peptides, metabolites, and actives.
  • Provides better LC/ESI-MS response than reversed-phase HPLC columns.
  • Offers direct compatibility with SPE solvents.
  • Delivers complementary selectivity.
  • Achieves higher sensitivity and lower detection limits in drug metabolism research.
  • Ensures universal compatibility with all LC detectors.
  • Guarantees excellent column-to-column repeatability.
  • Improves peak morphologies for polar bases.
  • Facilitates faster and easier method development.