Learn More
Waters Corp Atlantis HILIC Column, 100Å, 3 μm, 2.1 × 100 mm
Catalog No. 50780869 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Atlantis HILIC Silica Column is designed for use with a high percentage of organic mobile phase (greater than 80% acetonitrile) to retain analytes that are too polar to be held by reversed-phase chromatography. It offers orthogonal selectivity compared to conventional reversed-phase chromatography methods.
  • Can be used with a high percentage of organic mobile phase (>80% acetonitrile).
  • Retains analytes that are too polar to be retained by reversed-phase chromatography.
  • Provides orthogonal selectivity compared to traditional reversed-phase chromatography.
Catalog No. 50-780-869 Supplier Waters Corp Supplier No. 186002013
The Atlantis HILIC Silica Column is designed for use with a high percentage of organic mobile phase (greater than 80% acetonitrile) to retain analytes that are too polar to be held by reversed-phase chromatography. It offers orthogonal selectivity compared to conventional reversed-phase chromatography methods.
  • Can be used with a high percentage of organic mobile phase (>80% acetonitrile).
  • Retains analytes that are too polar to be retained by reversed-phase chromatography.
  • Provides orthogonal selectivity compared to traditional reversed-phase chromatography.