Learn More
Waters Corp Atlantis HILIC Column, 100Å, 5 µm, 4.6 mm X 100 mm, 1/pk
Catalog No. 50780886 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Atlantis HILIC Silica columns are used in combination with high organic mobile phases (>80% acetonitrile) to provide retention of analytes that are simply too polar to be retained by reverse-phase chromatography. HILIC separation mechanisms provide orthogonal analyte selectivity compared to traditional chromatographic reversed-phase approaches.
Catalog No. 50-780-886 Supplier Waters Corp Supplier No. 186002030
Atlantis HILIC Silica columns are used in combination with high organic mobile phases (>80% acetonitrile) to provide retention of analytes that are simply too polar to be retained by reverse-phase chromatography. HILIC separation mechanisms provide orthogonal analyte selectivity compared to traditional chromatographic reversed-phase approaches.