Learn More
Waters Corp HPLC Column, 100 Å, 3 μm, 2.1 mm x 15 mm, HILIC
Catalog No. 50780863 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Atlantis Silica HILIC Direct Connect Column is designed for Hydrophilic Interaction Chromatography (HILIC) separations in HPLC. It features a silica matrix with a 100 Å pore size and 3 μm spherical, fully porous particles. Constructed from robust stainless steel, this analytical column is ready-to-use for polar and hydrophilic analyte separation.
  • Optimized for HILIC separations of polar compounds
  • Silica chemistry with non-endcapping for HILIC retention
  • Robust stainless steel hardware for HPLC technology
  • Stable pH range of 1.0 to 5.0
  • High pressure stability up to 415 bar
  • Spherical, fully porous particles for efficient separation
  • Ideal for hydrophilic analyte separation, complementing reversed-phase chromatography
  • Supports HILIC mobile phases for enhanced polar compound retention
Catalog No. 50-780-863 Supplier Waters Corp Supplier No. 186002007
The Atlantis Silica HILIC Direct Connect Column is designed for Hydrophilic Interaction Chromatography (HILIC) separations in HPLC. It features a silica matrix with a 100 Å pore size and 3 μm spherical, fully porous particles. Constructed from robust stainless steel, this analytical column is ready-to-use for polar and hydrophilic analyte separation.
  • Optimized for HILIC separations of polar compounds
  • Silica chemistry with non-endcapping for HILIC retention
  • Robust stainless steel hardware for HPLC technology
  • Stable pH range of 1.0 to 5.0
  • High pressure stability up to 415 bar
  • Spherical, fully porous particles for efficient separation
  • Ideal for hydrophilic analyte separation, complementing reversed-phase chromatography
  • Supports HILIC mobile phases for enhanced polar compound retention