Learn More
Waters Corp Waters Sentry Guard Cartridge, Atlantis HILIC, Silica, 3 μm, 100 Å
Catalog No. 50780861 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Sentry Guard Cartridge is designed for HILIC Liquid Chromatography Columns. It extends analytical column life by removing chemical and particulate contamination, suitable for retaining analytes too polar for reverse-phase chromatography.
  • Used with high organic mobile phases for retaining analytes too polar for reverse-phase chromatography.
  • Offers orthogonal analyte selectivity.
  • Extends analytical column life by removing chemical and particulate contamination.
  • Ensures optimized performance with compatible HILIC Liquid Chromatography Columns.
  • Single unit that attaches directly to the column.
  • Manufactured under strict cGMP and ISO 9001 conditions.
  • Tested and certified to deliver optimal performance.
  • Maintains separation efficiency of UPLC, UHPLC, and HPLC.
Catalog No. 50-780-861 Supplier Waters Corp Supplier No. 186002005
This Sentry Guard Cartridge is designed for HILIC Liquid Chromatography Columns. It extends analytical column life by removing chemical and particulate contamination, suitable for retaining analytes too polar for reverse-phase chromatography.
  • Used with high organic mobile phases for retaining analytes too polar for reverse-phase chromatography.
  • Offers orthogonal analyte selectivity.
  • Extends analytical column life by removing chemical and particulate contamination.
  • Ensures optimized performance with compatible HILIC Liquid Chromatography Columns.
  • Single unit that attaches directly to the column.
  • Manufactured under strict cGMP and ISO 9001 conditions.
  • Tested and certified to deliver optimal performance.
  • Maintains separation efficiency of UPLC, UHPLC, and HPLC.