Learn More
Waters Corp Column, 5 μm, 4.6 x 150 mm, BEH C18 AX
Catalog No. 502391458 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
  • Excellent retention for polar acidic analytes under reversed phase conditions.
  • Reversed phase/anion-exchange mixed mode chemistry.
  • Stable from pH 2 to 10.
  • Complimentary selectivity to traditional C18 stationary phases.
  • Breakthrough Ethylene-Bridged Hybrid (BEH) particle technology for high retentivity and improved sample loadability.
  • MaxPeak HPS technology to prevent unwanted analyte-surface interactions.
  • Designed to prevent sample loss during transfer from HPLC to UHPLC or UPLC systems.
  • Employs a charge modifier for incredible retentivity for acids across a wide pH range.
Catalog No. 50-239-1458 Supplier Waters Corp Supplier No. 186009417
  • Excellent retention for polar acidic analytes under reversed phase conditions.
  • Reversed phase/anion-exchange mixed mode chemistry.
  • Stable from pH 2 to 10.
  • Complimentary selectivity to traditional C18 stationary phases.
  • Breakthrough Ethylene-Bridged Hybrid (BEH) particle technology for high retentivity and improved sample loadability.
  • MaxPeak HPS technology to prevent unwanted analyte-surface interactions.
  • Designed to prevent sample loss during transfer from HPLC to UHPLC or UPLC systems.
  • Employs a charge modifier for incredible retentivity for acids across a wide pH range.