Learn More
Waters Corp Column, Atlantis Premier BEH C18 AX, 5 μm, 4.6 x 100 mm
Catalog No. 502392466 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This reversed-phase/anion-exchange mixed mode column is designed for challenging liquid chromatography (LC) separations. It provides excellent retention for polar acidic analytes, complementary selectivity for method development, and utilizes Ethylene-Bridged Hybrid (BEH) particle technology for robust performance and improved resolution of complex glycans.
  • Offers excellent retention for polar acidic analytes.
  • Mixed-mode (RP/anion-exchange) chemistry stable from pH 2 to 10.
  • Offers complementary selectivity for method development & reproducibility.
  • Maximizes throughput for demanding LC separations.
  • Features Ethylene-Bridged Hybrid (BEH) particle technology.
  • Seamless method transfer to UPLC and UHPLC systems.
  • Designed for charge-based separation of neutral-to-highly acidic N-glycans.
  • Improves resolution and recovery for sialylated and phosphorylated glycans.
Catalog No. 50-239-2466 Supplier Waters Corp Supplier No. 186009416
This reversed-phase/anion-exchange mixed mode column is designed for challenging liquid chromatography (LC) separations. It provides excellent retention for polar acidic analytes, complementary selectivity for method development, and utilizes Ethylene-Bridged Hybrid (BEH) particle technology for robust performance and improved resolution of complex glycans.
  • Offers excellent retention for polar acidic analytes.
  • Mixed-mode (RP/anion-exchange) chemistry stable from pH 2 to 10.
  • Offers complementary selectivity for method development & reproducibility.
  • Maximizes throughput for demanding LC separations.
  • Features Ethylene-Bridged Hybrid (BEH) particle technology.
  • Seamless method transfer to UPLC and UHPLC systems.
  • Designed for charge-based separation of neutral-to-highly acidic N-glycans.
  • Improves resolution and recovery for sialylated and phosphorylated glycans.