Learn More
Waters Corp Waters Column, BEH C18 AX, 2.5 μm, 4.6 mm ID, 100 mm Length
Catalog No. 502039026 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Atlantis Premier BEH C18 AX Columns provide excellent retention for polar acidic analytes under reversed-phase conditions. The mixed-mode chemistry, combining reversed-phase and anion-exchange properties, is stable within a pH range of 2-10 and offers complementary selectivity to traditional C18 stationary phases, beneficial for method development.
  • Excellent retention for polar acidic analytes
  • Mixed-mode chemistry (reversed-phase/anion-exchange)
  • Stable within pH range of 2-10
  • Complementary selectivity to traditional C18 phases
  • Hybrid particle substrate
  • 2.5 μm particle size
  • 4.6 mm inner diameter
  • 100 mm length
  • Maximum pressure of 18000 psi (1240 bar)
  • Suitable for UPLC and UHPLC systems
Catalog No. 50-203-9026 Supplier Waters Corp Supplier No. 186009397
The Atlantis Premier BEH C18 AX Columns provide excellent retention for polar acidic analytes under reversed-phase conditions. The mixed-mode chemistry, combining reversed-phase and anion-exchange properties, is stable within a pH range of 2-10 and offers complementary selectivity to traditional C18 stationary phases, beneficial for method development.
  • Excellent retention for polar acidic analytes
  • Mixed-mode chemistry (reversed-phase/anion-exchange)
  • Stable within pH range of 2-10
  • Complementary selectivity to traditional C18 phases
  • Hybrid particle substrate
  • 2.5 μm particle size
  • 4.6 mm inner diameter
  • 100 mm length
  • Maximum pressure of 18000 psi (1240 bar)
  • Suitable for UPLC and UHPLC systems