Learn More
Waters Corp Atlantis C18 AX Method Validation Kit, 2.5 μm, 2.1 x 100 mm
Catalog No. 502392824 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Atlantis Premier BEH C18 AX Method Validation Kit contains three columns, each packed with a different lot of material. These columns provide excellent retention for polar acidic analytes under reversed-phase conditions. Featuring reversed-phase/anion-exchange mixed-mode chemistry, the kit is stable from pH 2 to 10 and offers complementary selectivity to traditional C18 stationary phases, beneficial for method development.
  • Contains three columns, each packed with a different lot of material.
  • Provides excellent retention for polar acidic analytes under reversed-phase conditions.
  • Reversed-phase/anion-exchange mixed-mode chemistry.
  • Stable from pH 2 to 10.
  • Offers complementary selectivity to traditional C18 stationary phases, beneficial for method development.
Catalog No. 50-239-2824 Supplier Waters Corp Supplier No. 186009395
The Atlantis Premier BEH C18 AX Method Validation Kit contains three columns, each packed with a different lot of material. These columns provide excellent retention for polar acidic analytes under reversed-phase conditions. Featuring reversed-phase/anion-exchange mixed-mode chemistry, the kit is stable from pH 2 to 10 and offers complementary selectivity to traditional C18 stationary phases, beneficial for method development.
  • Contains three columns, each packed with a different lot of material.
  • Provides excellent retention for polar acidic analytes under reversed-phase conditions.
  • Reversed-phase/anion-exchange mixed-mode chemistry.
  • Stable from pH 2 to 10.
  • Offers complementary selectivity to traditional C18 stationary phases, beneficial for method development.