Learn More
Waters Corp Method Validation Kit, BEH C18 AX, 1.7 μm, VanGuard FIT
Catalog No. 502391226 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This method validation kit provides three columns, each packed with a distinct lot of material. It offers excellent retention for polar acidic analytes under reversed-phase conditions with mixed-mode chemistry (reversed-phase/anion-exchange) stable from pH 2 to 10. The VanGuard FIT column format minimizes dead volume and extends column lifetime.
  • Kit contains three columns, each packed with a different lot of material.
  • Provides excellent retention for polar acidic analytes under reversed-phase conditions.
  • Mixed-mode chemistry is stable from pH 2 to 10.
  • Offers complementary selectivity to traditional C18 stationary phases for method development.
  • VanGuard FIT column format reduces dead volume and increases column lifetime.
Catalog No. 50-239-1226 Supplier Waters Corp Supplier No. 186009363
This method validation kit provides three columns, each packed with a distinct lot of material. It offers excellent retention for polar acidic analytes under reversed-phase conditions with mixed-mode chemistry (reversed-phase/anion-exchange) stable from pH 2 to 10. The VanGuard FIT column format minimizes dead volume and extends column lifetime.
  • Kit contains three columns, each packed with a different lot of material.
  • Provides excellent retention for polar acidic analytes under reversed-phase conditions.
  • Mixed-mode chemistry is stable from pH 2 to 10.
  • Offers complementary selectivity to traditional C18 stationary phases for method development.
  • VanGuard FIT column format reduces dead volume and increases column lifetime.