Learn More
Waters Corp Column, Atlantis Premier BEH C18 AX VanGuard FIT, 5 μm, 2.1 x 50 mm
Catalog No. 502392294 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Atlantis Premier BEH C18 AX Columns provide excellent retention for polar acidic analytes with stable reversed-phase/anion-exchange mixed-mode chemistry (pH 2 to 10). The VanGuard FIT guard-column design reduces dead volume and increases column lifetime by preventing contaminants from reaching the analytical column.
  • Retains polar acidic analytes under reversed-phase conditions.
  • Stable reversed-phase/anion-exchange mixed-mode chemistry (pH 2 to 10).
  • VanGuard FIT guard-column design reduces dead volume and extends column lifetime.
  • Protects analytical column from contaminants like sample matrix and particles.
  • Constructed with 5 μm particle size, 2.1 x 50 mm dimensions.
  • Ideal for biological fluid cleanup and temperature-controlled applications.
  • Operates at pH 10 max, 60°C max, and 18,000 psi (1240 Bar) max pressure.
Catalog No. 50-239-2294 Supplier Waters Corp Supplier No. 186009404
The Atlantis Premier BEH C18 AX Columns provide excellent retention for polar acidic analytes with stable reversed-phase/anion-exchange mixed-mode chemistry (pH 2 to 10). The VanGuard FIT guard-column design reduces dead volume and increases column lifetime by preventing contaminants from reaching the analytical column.
  • Retains polar acidic analytes under reversed-phase conditions.
  • Stable reversed-phase/anion-exchange mixed-mode chemistry (pH 2 to 10).
  • VanGuard FIT guard-column design reduces dead volume and extends column lifetime.
  • Protects analytical column from contaminants like sample matrix and particles.
  • Constructed with 5 μm particle size, 2.1 x 50 mm dimensions.
  • Ideal for biological fluid cleanup and temperature-controlled applications.
  • Operates at pH 10 max, 60°C max, and 18,000 psi (1240 Bar) max pressure.