Learn More
Waters Corp Atlantis Method Validation Kit, Premier BEH C18 AX VanGuard FIT, 2.5 μm, 4.6 x 150 mm, 3/pk
Supplier:  Waters Corp 186009388
Encompass_Preferred
  • Includes three columns, each from a different material lot.
  • Excellent retention for polar acidic analytes in reversed-phase conditions.
  • Reversed-phase/anion-exchange mixed-mode chemistry offers pH 2 to 10 stability.
  • Provides complementary selectivity for method development.
  • All-in-one guard-column design minimizes dead volume.
  • Extends column lifetime.
  • Hybrid spherical particle substrate.
  • Low silanol activity.
  • Compatible with UPLC, UHPLC, and LC techniques.
  • Meets USP classification L78.
Catalog No. 50-239-2760