Learn More
Waters Corp Premier BEH C18 AX VanGuard FIT Column, 5 μm, 4.6 x 100 mm
Supplier:  Waters Corp 186009411
Encompass_Preferred
The Premier BEH C18 AX Columns offer excellent retention for polar acidic analytes with a stable reversed phase/anion-exchange mixed mode chemistry. Its VanGuard FIT design reduces dead volume and extends column lifetime for reliable HPLC analysis.
  • Excellent retention for polar acidic analytes.
  • Stable RP/anion-exchange mixed mode chemistry (pH 2–10).
  • VanGuard FIT design reduces dead volume, extends column life.
  • BEH particle technology for precise analytical outcomes.
  • 5 μm particle size, 4.6 mm ID, 100 mm length.
  • Protects main column from contamination.
  • Applications: amino acid, peptide, pharmaceutical, environmental analysis.
  • Includes performance chromatogram and batch analysis certificate.
Catalog No. 50-239-2870