Learn More
Waters Corp Column, 5 μm, 4.6 x 250 mm, BEH C18 AX VanGuard FIT
Catalog No. 502393023 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Atlantis Premier BEH C18 AX Columns offer excellent retention for polar acidic analytes with mixed-mode chemistry, providing complementary selectivity and pH 2–10 stability. The VanGuard FIT Column reduces dead volume and extends column life by protecting against sample contaminants. MaxPeak High Performance Surfaces (HPS) technology minimizes analyte/surface interactions, improving recovery and sensitivity.
  • Excellent retention for polar acidic analytes.
  • Stable from pH 2–10.
  • Mixed-mode chemistry offers complementary selectivity.
  • VanGuard FIT Column reduces dead volume and extends column life.
  • Protects column from contaminants.
  • MaxPeak HPS technology improves recovery, sensitivity, and reproducibility.
  • Reduces conditioning and passivation times.
  • Manufactured in a cGMP, ISO 9001 certified plant.
Catalog No. 50-239-3023 Supplier Waters Corp Supplier No. 186009413
The Atlantis Premier BEH C18 AX Columns offer excellent retention for polar acidic analytes with mixed-mode chemistry, providing complementary selectivity and pH 2–10 stability. The VanGuard FIT Column reduces dead volume and extends column life by protecting against sample contaminants. MaxPeak High Performance Surfaces (HPS) technology minimizes analyte/surface interactions, improving recovery and sensitivity.
  • Excellent retention for polar acidic analytes.
  • Stable from pH 2–10.
  • Mixed-mode chemistry offers complementary selectivity.
  • VanGuard FIT Column reduces dead volume and extends column life.
  • Protects column from contaminants.
  • MaxPeak HPS technology improves recovery, sensitivity, and reproducibility.
  • Reduces conditioning and passivation times.
  • Manufactured in a cGMP, ISO 9001 certified plant.