Learn More
Waters Corp (H)PLC Column, Atlantis PREMIER BEH C18 AX, 95 Å, 2.5 μm
Catalog No. 502391720 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This (H)PLC Column Atlantis PREMIER BEH C18 AX features a 95 Å pore size, 2.5 μm particle size, and measures 2.1 × 50 mm. It is a VanGuard FIT Method Validation Kit (MVK) and comes in a 3 piece per pack configuration.
  • Product-specific features and benefits were not found in the provided research data.
Catalog No. 50-239-1720 Supplier Waters Corp Supplier No. 186009380
Add to Cart
Add to Cart
This (H)PLC Column Atlantis PREMIER BEH C18 AX features a 95 Å pore size, 2.5 μm particle size, and measures 2.1 × 50 mm. It is a VanGuard FIT Method Validation Kit (MVK) and comes in a 3 piece per pack configuration.
  • Product-specific features and benefits were not found in the provided research data.