Learn More
Waters Corp Waters Column, RP/Anion-exchange, 1.7 μm, 2.1 x 100 mm
Catalog No. 502038997 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This column excels at retaining polar acidic analytes in reversed-phase conditions. Its stable reversed-phase/anion-exchange mixed-mode chemistry, effective from pH 2–10, offers unique selectivity compared to traditional C18 phases for method development. The VanGuard FIT Column format is an all-in-one guard-column designed to reduce dead volume and extend column lifetime by filtering impurities.
  • Excellent retention for polar acidic analytes under reversed phase conditions.
  • Stable reversed phase/anion-exchange mixed mode chemistry from pH 2–10 for method development.
  • All-in-one guard-column design reduces dead volume and increases column lifetime.
  • Removes chemical impurities and suspended solids, enhancing column reliability.
  • Tolerates pressures up to 15000 psi (1030 bar) in UPLC System applications.
Catalog No. 50-203-8997 Supplier Waters Corp Supplier No. 186009360
This column excels at retaining polar acidic analytes in reversed-phase conditions. Its stable reversed-phase/anion-exchange mixed-mode chemistry, effective from pH 2–10, offers unique selectivity compared to traditional C18 phases for method development. The VanGuard FIT Column format is an all-in-one guard-column designed to reduce dead volume and extend column lifetime by filtering impurities.
  • Excellent retention for polar acidic analytes under reversed phase conditions.
  • Stable reversed phase/anion-exchange mixed mode chemistry from pH 2–10 for method development.
  • All-in-one guard-column design reduces dead volume and increases column lifetime.
  • Removes chemical impurities and suspended solids, enhancing column reliability.
  • Tolerates pressures up to 15000 psi (1030 bar) in UPLC System applications.