Learn More
Waters Corp Waters Column, BEH C18 AX VanGuard FIT, 1.7 μm, 2.1 x 150 mm
Catalog No. NC1821383 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This column provides excellent retention for polar acidic analytes under reversed-phase conditions. Its mixed-mode chemistry is stable from pH 2–10, offering complementary selectivity. The VanGuard FIT Column format, an all-in-one guard-column design, reduces dead volume and increases column lifetime, protecting the column from unwanted particles and boosting performance.
  • Excellent retention for polar acidic analytes in reversed-phase.
  • Mixed mode chemistry, stable from pH 2–10.
  • Complimentary selectivity to traditional C18 phases.
  • VanGuard FIT Column, all-in-one guard-column design.
  • Reduces dead volume, increases column lifetime.
  • Manufactured with high-performance packing materials.
  • Suitable for biological fluid cleanup and temperature control.
  • Protects column from unwanted sample matrix or particles.
  • Boosts and improves analytical performance.
Catalog No. NC1821383 Supplier Waters Corp Supplier No. 186009361
This column provides excellent retention for polar acidic analytes under reversed-phase conditions. Its mixed-mode chemistry is stable from pH 2–10, offering complementary selectivity. The VanGuard FIT Column format, an all-in-one guard-column design, reduces dead volume and increases column lifetime, protecting the column from unwanted particles and boosting performance.
  • Excellent retention for polar acidic analytes in reversed-phase.
  • Mixed mode chemistry, stable from pH 2–10.
  • Complimentary selectivity to traditional C18 phases.
  • VanGuard FIT Column, all-in-one guard-column design.
  • Reduces dead volume, increases column lifetime.
  • Manufactured with high-performance packing materials.
  • Suitable for biological fluid cleanup and temperature control.
  • Protects column from unwanted sample matrix or particles.
  • Boosts and improves analytical performance.