Learn More
Waters Corp Waters Column, BEH Z-HILIC, Sulfobetaine, 1.7 μm, 2.1 x 150 mm
Catalog No. 502224382 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Atlantis Premier BEH Z-HILIC Columns are designed to enhance polar analyte retention and provide distinct selectivity compared to other HILIC chemistries, making them valuable for HILIC methods development. These columns feature robust ethylene bridged hybrid particles, ensuring pH stability from pH 2 to 10. MaxPeak High Performance Surface hardware technology minimizes unwanted analyte/surface interactions, preventing poor peak shape and signal intensity loss.
  • Increase polar analyte retention and offer different selectivity for HILIC methods development.
  • Constructed with rugged ethylene bridged hybrid particles, ensuring pH stability from pH 2 to 10.
  • MaxPeak High Performance Surface hardware technology reduces unwanted analyte/surface interactions, addressing issues like poor peak shape and signal intensity losses.
This item is not returnable. View return policy
Catalog No. 50-222-4382 Supplier Waters Corp Supplier No. 186009980
Atlantis Premier BEH Z-HILIC Columns are designed to enhance polar analyte retention and provide distinct selectivity compared to other HILIC chemistries, making them valuable for HILIC methods development. These columns feature robust ethylene bridged hybrid particles, ensuring pH stability from pH 2 to 10. MaxPeak High Performance Surface hardware technology minimizes unwanted analyte/surface interactions, preventing poor peak shape and signal intensity loss.
  • Increase polar analyte retention and offer different selectivity for HILIC methods development.
  • Constructed with rugged ethylene bridged hybrid particles, ensuring pH stability from pH 2 to 10.
  • MaxPeak High Performance Surface hardware technology reduces unwanted analyte/surface interactions, addressing issues like poor peak shape and signal intensity losses.