Learn More
Waters Corp Waters Column, 5 μm, 4.6 mm x 250 mm, BEH Z-HILIC
Supplier:  Waters Corp 186010009
Encompass_Preferred
Atlantis Premier BEH Z-HILIC columns are designed for hydrophilic interaction liquid chromatography (HILIC). They feature MaxPeak High Performance Surface technology to minimize unwanted analyte interactions, providing enhanced polar analyte retention, distinct selectivity, and a wide pH stability range.
  • Enhances polar analyte retention and offers distinct selectivity for HILIC methods.
  • Constructed with durable ethylene bridged hybrid particles for pH stability from pH 2 to 10.
  • MaxPeak High Performance Surface hardware technology reduces unwanted analyte/surface interactions.
  • Reduces issues like poor peak shape and loss in signal intensity caused by analyte interactions.
  • Utilizes sulfobetaine zwitterionic chemistry for high retention of polar neutral compounds.
  • Provides alternative selectivity for polar acidic and basic compounds.
Catalog No. 50-222-4411