Learn More
Waters Corp Waters Column, BEH Z-HILIC, 2.5 μm, 2.1 mm x 100 mm
Supplier:  Waters Corp 186009986
Encompass_Preferred
Atlantis Premier BEH Z-HILIC Columns enhance polar analyte retention and offer distinct selectivity for HILIC methods development. Built with robust ethylene bridged hybrid particles, they ensure stability across a pH range of 2–10. MaxPeak High Performance Surface technology significantly reduces unwanted analyte/surface interactions, improving peak shape and signal intensity.
  • Enhances polar analyte retention and offers distinct selectivity
  • Constructed with rugged ethylene bridged hybrid particles
  • Stable across a wide pH range of 2–10
  • MaxPeak High Performance Surface technology minimizes analyte/surface interactions
  • Compatible with UHPLC and UPLC systems; features sulfobetaine zwitterionic chemistry
  • Low silanol activity, spherical particle shape, and 95 Å pore size
Catalog No. 50-222-4388