Learn More
Waters Corp Waters Corporation Column, Atlantis Premier BEH Z-HILIC, 1.7 μm, 2.1 mm x 100 mm
Catalog No. 502224384 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
  • Provides an increase in polar analyte retention.
  • Offers a different selectivity when compared to other HILIC chemistries.
  • Built using rugged ethylene bridged hybrid particles.
  • pH stable from pH 2 to 10.
  • MaxPeak High Performance Surface hardware technology significantly reduces unwanted analyte/surface interactions.
  • VanGuard FIT column format is an all-in-one, guard-column design.
  • Engineered to reduce dead volume.
  • Engineered to increase column lifetime.
Catalog No. 50-222-4384 Supplier Waters Corp Supplier No. 186009982
  • Provides an increase in polar analyte retention.
  • Offers a different selectivity when compared to other HILIC chemistries.
  • Built using rugged ethylene bridged hybrid particles.
  • pH stable from pH 2 to 10.
  • MaxPeak High Performance Surface hardware technology significantly reduces unwanted analyte/surface interactions.
  • VanGuard FIT column format is an all-in-one, guard-column design.
  • Engineered to reduce dead volume.
  • Engineered to increase column lifetime.