Learn More
Waters Corp BEH Z-HILIC Column, 2.5 μm, 2.1 x 100 mm
Catalog No. NC3353203 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Atlantis Premier BEH Z-HILIC columns offer increased polar analyte retention and unique selectivity for HILIC methods. They are built with rugged ethylene bridged hybrid particles, ensuring pH stability from pH 2 to 10. MaxPeak High Performance Surface technology reduces unwanted analyte/surface interactions, preventing poor peak shape and signal loss. The VanGuard FIT column format is an all-in-one guard-column, designed to minimize dead volume and extend column lifetime.
  • Increased polar analyte retention and different selectivity for HILIC methods.
  • Rugged ethylene bridged hybrid particles provide pH stability from pH 2 to 10.
  • MaxPeak High Performance Surface technology reduces unwanted analyte/surface interactions, improving peak shape and signal.
  • VanGuard FIT all-in-one guard-column minimizes dead volume and extends column lifetime.
Catalog No. NC3353203 Supplier Waters Corp Supplier No. 186009989
Atlantis Premier BEH Z-HILIC columns offer increased polar analyte retention and unique selectivity for HILIC methods. They are built with rugged ethylene bridged hybrid particles, ensuring pH stability from pH 2 to 10. MaxPeak High Performance Surface technology reduces unwanted analyte/surface interactions, preventing poor peak shape and signal loss. The VanGuard FIT column format is an all-in-one guard-column, designed to minimize dead volume and extend column lifetime.
  • Increased polar analyte retention and different selectivity for HILIC methods.
  • Rugged ethylene bridged hybrid particles provide pH stability from pH 2 to 10.
  • MaxPeak High Performance Surface technology reduces unwanted analyte/surface interactions, improving peak shape and signal.
  • VanGuard FIT all-in-one guard-column minimizes dead volume and extends column lifetime.